3-[(4-Aminophenyl)thio]-1-(5-methylisoxazol-3-yl)pyrrolidine-2,5-dione

Supplier: Matrix Scientific

038884-500MG
101846-404EA 144.9 USD
101846-404
3-[(4-Aminophenyl)thio]-1-(5-methylisoxazol-3-yl)pyrrolidine-2,5-dione
3-[(4-Aminophenyl)thio]-1-(5-methylisoxazol-3-yl)pyrrolidine-2,5-dione

Hazard: Irritant

Formula: C₁₄H₁₃N₃O₃S
MDL Number: MFCD09669366
CAS Number: 931291-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR