3-[(4-Aminophenyl)thio]-1-(2,6-dichlorophenyl)pyrrolidine-2,5-dione

Supplier: Matrix Scientific

038882-500MG
101846-400EA 144.9 USD
101846-400
3-[(4-Aminophenyl)thio]-1-(2,6-dichlorophenyl)pyrrolidine-2,5-dione
3-[(4-Aminophenyl)thio]-1-(2,6-dichlorophenyl)pyrrolidine-2,5-dione

Hazard: Irritant

Formula: C₁₆H₁₂Cl₂N₂O₂S
MDL Number: MFCD09669225
CAS Number: 931259-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR