5-Methoxy-1-methyl-1H-indole-2-carbohydrazide

Supplier: Matrix Scientific

038874-500MG
101846-380EA 217.35 USD
101846-380
5-Methoxy-1-methyl-1H-indole-2-carbohydrazide
5-Methoxy-1-methyl-1H-indole-2-carbohydrazide

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₂
MDL Number: MFCD00449859
CAS Number: 200062-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR