4,6,7-Trimethoxy-1H-indole-2-carbohydrazide

Supplier: Matrix Scientific

038871-500MG
101846-374EA 217.35 USD
101846-374
4,6,7-Trimethoxy-1H-indole-2-carbohydrazide
4,6,7-Trimethoxy-1H-indole-2-carbohydrazide

Hazard: Irritant

Formula: C₁₂H₁₅N₃O₄
MDL Number: MFCD12027496
CAS Number: 1134334-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR