(5-Ethoxy-2-methyl-1H-indol-3-yl)acetic acid

Supplier: Matrix Scientific

038868-500MG
101846-368EA 217.35 USD
101846-368
(5-Ethoxy-2-methyl-1H-indol-3-yl)acetic acid
(5-Ethoxy-2-methyl-1H-indol-3-yl)acetic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₃
MDL Number: MFCD02664397
CAS Number: 34024-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR