3-Bromo-2-methyl-1-(2-methyl-1H-indol-3-yl)propan-1-one

Supplier: Matrix Scientific

038862-500MG
101846-356EA 217.35 USD
101846-356
3-Bromo-2-methyl-1-(2-methyl-1H-indol-3-yl)propan-1-one
3-Bromo-2-methyl-1-(2-methyl-1H-indol-3-yl)propan-1-one

Hazard: Irritant

Formula: C₁₃H₁₄BrNO
MDL Number: MFCD12027492
CAS Number: 1134334-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR