3-Bromo-1-(5-methoxy-1H-indol-3-yl)-2-methylpropan-1-one

Supplier: Matrix Scientific

038860-500MG
101846-352EA 217.35 USD
101846-352
3-Bromo-1-(5-methoxy-1H-indol-3-yl)-2-methylpropan-1-one
3-Bromo-1-(5-methoxy-1H-indol-3-yl)-2-methylpropan-1-one

Hazard: Irritant

Formula: C₁₃H₁₄BrNO₂
MDL Number: MFCD12027490
CAS Number: 1134334-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR