2-Chloro-1-(6-methyl-1H-indol-3-yl)ethanone

Supplier: Matrix Scientific

038853-500MG
101846-338EA 217.35 USD
101846-338
2-Chloro-1-(6-methyl-1H-indol-3-yl)ethanone
2-Chloro-1-(6-methyl-1H-indol-3-yl)ethanone

Hazard: Irritant

Formula: C₁₁H₁₀ClNO
MDL Number: MFCD03848185
CAS Number: 115027-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR