2-Chloro-1-(2,7-dimethyl-1H-indol-3-yl)ethanone

Supplier: Matrix Scientific

038851-500MG
101846-334EA 217.35 USD
101846-334
2-Chloro-1-(2,7-dimethyl-1H-indol-3-yl)ethanone
2-Chloro-1-(2,7-dimethyl-1H-indol-3-yl)ethanone

Hazard: Irritant

Formula: C₁₂H₁₂ClNO
MDL Number: MFCD12027484
CAS Number: 1134334-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR