2-Chloro-1-(5-chloro-1H-indol-3-yl)ethanone

Supplier: Matrix Scientific

038849-500MG
101846-330EA 217.35 USD
101846-330
2-Chloro-1-(5-chloro-1H-indol-3-yl)ethanone
2-Chloro-1-(5-chloro-1H-indol-3-yl)ethanone

Hazard: Irritant

Formula: C₁₀H₇Cl₂NO
MDL Number: MFCD03848174
CAS Number: 38693-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR