Methyl-5-chloro-3-[(chloroacetyl)amino]-1H-indole-2-carboxylate

Supplier: Matrix Scientific

038843-500MG
101846-318EA 217.35 USD
101846-318
Methyl-5-chloro-3-[(chloroacetyl)amino]-1H-indole-2-carboxylate
Methyl-5-chloro-3-[(chloroacetyl)amino]-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₀Cl₂N₂O₃
MDL Number: MFCD12027479
CAS Number: 1134334-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR