Ethyl-3-[(chloroacetyl)amino]-5-fluoro-1H-indole-2-carboxylate

Supplier: Matrix Scientific

038841-500MG
101846-314EA 290.77 USD
101846-314
Ethyl-3-[(chloroacetyl)amino]-5-fluoro-1H-indole-2-carboxylate
Ethyl-3-[(chloroacetyl)amino]-5-fluoro-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₂ClFN₂O₃
MDL Number: MFCD12027477
CAS Number: 1134334-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR