Methyl-3-[(chloroacetyl)amino]-6-methoxy-1H-indole-2-carboxylate

Supplier: Matrix Scientific

038839-500MG
101846-310EA 217.35 USD
101846-310
Methyl-3-[(chloroacetyl)amino]-6-methoxy-1H-indole-2-carboxylate
Methyl-3-[(chloroacetyl)amino]-6-methoxy-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₃ClN₂O₄
MDL Number: MFCD03848214
CAS Number: 843638-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR