4-Chloro-5,8-dimethyl-5H-pyrimido[5,4-b]indole

Supplier: Matrix Scientific

038837-500MG
101846-306EA 217.35 USD
101846-306
4-Chloro-5,8-dimethyl-5H-pyrimido[5,4-b]indole
4-Chloro-5,8-dimethyl-5H-pyrimido[5,4-b]indole

Hazard: Irritant

Formula: C₁₂H₁₀ClN₃
MDL Number: MFCD12027476
CAS Number: 1134334-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR