N,N-Dimethyl-1-(5-methyl-1H-indol-3-yl)methanamine

Supplier: Matrix Scientific

Synonyms: 5-Methylgramine

038824-500MG
101846-280EA 289.8 USD
101846-280
N,N-Dimethyl-1-(5-methyl-1H-indol-3-yl)methanamine
N,N-Dimethyl-1-(5-methyl-1H-indol-3-yl)methanamine

Hazard: Irritant

Formula: C₁₂H₁₆N₂
MW: 188.27 g/mol
MDL Number: MFCD00022732
CAS Number: 30218-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR