[(5-Methoxy-1,2-dimethyl-1H-indol-3-yl)methyl]dimethylamine

Supplier: Matrix Scientific

038823-500MG
101846-278EA 289.8 USD
101846-278
[(5-Methoxy-1,2-dimethyl-1H-indol-3-yl)methyl]dimethylamine
[(5-Methoxy-1,2-dimethyl-1H-indol-3-yl)methyl]dimethylamine

Hazard: Irritant

Formula: C₁₄H₂₀N₂O
MDL Number: MFCD12027467
CAS Number: 93865-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR