5-(6-Methoxy-1H-indol-3-yl)-1,3-thiazol-2-amine

Supplier: Matrix Scientific

038819-500MG
101846-270EA 289.8 USD
101846-270
5-(6-Methoxy-1H-indol-3-yl)-1,3-thiazol-2-amine
5-(6-Methoxy-1H-indol-3-yl)-1,3-thiazol-2-amine

Hazard: Irritant

Formula: C₁₂H₁₁N₃OS
MDL Number: MFCD12027464
CAS Number: 1134334-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR