4-Chloro-7-methoxy-5H-pyrimido[5,4-b]indole

Supplier: Matrix Scientific

038818-500MG
101846-268EA 217.35 USD
101846-268
4-Chloro-7-methoxy-5H-pyrimido[5,4-b]indole
4-Chloro-7-methoxy-5H-pyrimido[5,4-b]indole

Hazard: Irritant

Formula: C₁₁H₈ClN₃O
MDL Number: MFCD03848207
CAS Number: 676602-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR