7-Fluoro-2-methyl-2,3,4,9-tetrahydro-1H-β-carboline
Supplier: Matrix Scientific
038814-500MG
101846-260EA
289.8
USD
101846-260
7-Fluoro-2-methyl-2,3,4,9-tetrahydro-1H-β-carboline
7-Fluoro-2-methyl-2,3,4,9-tetrahydro-1H-beta-carboline
Hazard: Irritant
Formula:
C₁₂H₁₃FN₂ |
MDL Number:
MFCD12027463 CAS Number: 1134334-47-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...