5-Hydroxy-2-methyl-1H-indole-3-carbaldehyde

Supplier: Matrix Scientific

038807-500MG
101846-246EA 217.35 USD
101846-246
5-Hydroxy-2-methyl-1H-indole-3-carbaldehyde
5-Hydroxy-2-methyl-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₀H₉NO₂
MDL Number: MFCD12027461
CAS Number: 412021-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR