5-Fluoro-1-propyl-1H-indole-3-carbaldehyde

Supplier: Matrix Scientific

038800-500MG
101846-232EA 289.8 USD
101846-232
5-Fluoro-1-propyl-1H-indole-3-carbaldehyde
5-Fluoro-1-propyl-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₁₂FNO
MDL Number: MFCD12027454
CAS Number: 944465-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR