6-Methyl-1-propyl-1H-indole-3-carbaldehyde

Supplier: Matrix Scientific

038798-500MG
101846-228EA 144.9 USD
101846-228
6-Methyl-1-propyl-1H-indole-3-carbaldehyde
6-Methyl-1-propyl-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₅NO
MDL Number: MFCD12027452
CAS Number: 1134334-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR