1-Ethyl-5-fluoro-1H-indole-3-carbaldehyde

Supplier: Matrix Scientific

038793-500MG
101846-218EA 289.8 USD
101846-218
1-Ethyl-5-fluoro-1H-indole-3-carbaldehyde
1-Ethyl-5-fluoro-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₁H₁₀FNO
MDL Number: MFCD12027450
CAS Number: 620175-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR