Methyl-3-amino-6-methoxy-1H-indole-2-carboxylate

Supplier: Matrix Scientific

038789-500MG
101846-212EA 250.7 USD
101846-212
Methyl-3-amino-6-methoxy-1H-indole-2-carboxylate
Methyl-3-amino-6-methoxy-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
MDL Number: MFCD03848203
CAS Number: 696649-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR