Methyl-3-amino-5-chloro-1H-indole-2-carboxylate

Supplier: Matrix Scientific

038786-500MG
101846-208EA 217.35 USD
101846-208
Methyl-3-amino-5-chloro-1H-indole-2-carboxylate
Methyl-3-amino-5-chloro-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₉ClN₂O₂
MDL Number: MFCD07364723
CAS Number: 889950-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR