Methyl-3-(4-chlorophenyl)-1H-indole-2-carboxylate

Supplier: Matrix Scientific

038779-500MG
101846-194EA 217.35 USD
101846-194
Methyl-3-(4-chlorophenyl)-1H-indole-2-carboxylate
Methyl-3-(4-chlorophenyl)-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₆H₁₂ClNO₂
MDL Number: MFCD12027448
CAS Number: 1134334-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR