(2,4,7-Trimethyl-1H-indol-3-yl)acetic acid

Supplier: Matrix Scientific

038776-500MG
101846-188EA 217.35 USD
101846-188
(2,4,7-Trimethyl-1H-indol-3-yl)acetic acid
(2,4,7-Trimethyl-1H-indol-3-yl)acetic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₂
MDL Number: MFCD02664368
CAS Number: 5435-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR