4-Fluoro-1H-indole-2-carboxylic acid

Supplier: Matrix Scientific

038775-500MG
101846-186EA 65.55 USD
101846-186
4-Fluoro-1H-indole-2-carboxylic acid
4-Fluoro-1H-indole-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₆FNO₂
MW: 179.1500 g/mol
MDL Number: MFCD02664471
CAS Number: 399-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR