5,6-Dimethoxy-1H-indole-2-carboxylic acid

Supplier: Matrix Scientific

038772-500MG
101846-180EA 88.55 USD
101846-180
5,6-Dimethoxy-1H-indole-2-carboxylic acid
5,6-Dimethoxy-1H-indole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₁NO₄
MW: 221.21 g/mol
MDL Number: MFCD00015459
CAS Number: 88210-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR