1,6-Dimethyl-1H-indole-2-carboxylic acid

Supplier: Matrix Scientific

038771-500MG
101846-178EA 144.9 USD
101846-178
1,6-Dimethyl-1H-indole-2-carboxylic acid
1,6-Dimethyl-1H-indole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₁NO₂
MDL Number: MFCD07364696
CAS Number: 893730-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR