6-Bromo-1-methyl-1H-indole-2-carboxylic acid

Supplier: Matrix Scientific

038770-500MG
101846-176EA 217.35 USD
101846-176
6-Bromo-1-methyl-1H-indole-2-carboxylic acid
6-Bromo-1-methyl-1H-indole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₈BrNO₂
MDL Number: MFCD07364654
CAS Number: 885121-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR