Methyl-5,6-dimethoxy-1-methyl-1H-indole-2-carboxylate

Supplier: Matrix Scientific

038768-500MG
101846-172EA 217.35 USD
101846-172
Methyl-5,6-dimethoxy-1-methyl-1H-indole-2-carboxylate
Methyl-5,6-dimethoxy-1-methyl-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₅NO₄
MDL Number: MFCD07364632
CAS Number: 32045-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR