Ethyl-5-(benzyloxy)-1H-indole-2-carboxylate

Supplier: Matrix Scientific

Synonyms: 5-Benzyloxyindole-2-carboxylic acid ethyl ester

038767-500MG
101846-170EA 144.9 USD
101846-170
Ethyl-5-(benzyloxy)-1H-indole-2-carboxylate
Ethyl-5-(benzyloxy)-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₈H₁₇NO₃
MW: 295.34 g/mol
MDL Number: MFCD00022702
CAS Number: 37033-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR