Methyl-4-fluoro-1H-indole-2-carboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 4-fluoro-1H-indole-2-carboxylate

038766-500MG
101846-168EA 131.1 USD
101846-168
Methyl-4-fluoro-1H-indole-2-carboxylate
Methyl-4-fluoro-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₈FNO₂
MW: 193.18 g/mol
MDL Number: MFCD06653401
CAS Number: 113162-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR