4-(2-Mercapto-4,4-dimethylpyrimidin-1(4H)-yl)benzenesulfonamide

Supplier: Matrix Scientific

038756-500MG
101846-148EA 144.9 USD
101846-148
4-(2-Mercapto-4,4-dimethylpyrimidin-1(4H)-yl)benzenesulfonamide
4-(2-Mercapto-4,4-dimethylpyrimidin-1(4H)-yl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₂H₁₅N₃O₂S₂
MDL Number: MFCD12027443
CAS Number: 1142213-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR