3-(2-Mercapto-4,4-dimethylpyrimidin-1(4H)-yl)benzoic acid

Supplier: Matrix Scientific

038752-500MG
101846-140EA 144.9 USD
101846-140
3-(2-Mercapto-4,4-dimethylpyrimidin-1(4H)-yl)benzoic acid
3-(2-Mercapto-4,4-dimethylpyrimidin-1(4H)-yl)benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₂S
MDL Number: MFCD12027439
CAS Number: 1142213-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR