1-(4-Butoxyphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Supplier: Matrix Scientific

038748-500MG
101846-132EA 144.9 USD
101846-132
1-(4-Butoxyphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol
1-(4-Butoxyphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Hazard: Irritant

Formula: C₁₆H₂₂N₂OS
MDL Number: MFCD12027435
CAS Number: 1142213-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR