4,4-Dimethyl-1-(4-phenoxyphenyl)-1,4-dihydropyrimidine-2-thiol

Supplier: Matrix Scientific

038747-500MG
101846-130EA 144.9 USD
101846-130
4,4-Dimethyl-1-(4-phenoxyphenyl)-1,4-dihydropyrimidine-2-thiol
4,4-Dimethyl-1-(4-phenoxyphenyl)-1,4-dihydropyrimidine-2-thiol

Hazard: Irritant

Formula: C₁₈H₁₈N₂OS
MDL Number: MFCD12027434
CAS Number: 1142212-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR