1-(4-Ethoxyphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Supplier: Matrix Scientific

038746-500MG
101846-128EA 144.9 USD
101846-128
1-(4-Ethoxyphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol
1-(4-Ethoxyphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Hazard: Irritant

Formula: C₁₄H₁₈N₂OS
MDL Number: MFCD12027433
CAS Number: 1142212-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR