Methyl-2-(2-mercapto-4,4-dimethylpyrimidin-1(4H)-yl)benzoate

Supplier: Matrix Scientific

038742-500MG
101846-120EA 144.9 USD
101846-120
Methyl-2-(2-mercapto-4,4-dimethylpyrimidin-1(4H)-yl)benzoate
Methyl-2-(2-mercapto-4,4-dimethylpyrimidin-1(4H)-yl)benzoate

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₂S
MDL Number: MFCD12027429
CAS Number: 1142212-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR