1-(4-Fluorophenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Supplier: Matrix Scientific

038740-500MG
101846-116EA 144.9 USD
101846-116
1-(4-Fluorophenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol
1-(4-Fluorophenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Hazard: Irritant

Formula: C₁₂H₁₃FN₂S
MDL Number: MFCD12027427
CAS Number: 1142212-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR