Methyl-6-cyano-1H-indole-2-carboxylate
Supplier: Matrix Scientific
Synonyms:
Methyl 6-cyano-1H-indole-2-carboxylate
038730-500MG
101846-096EA
230.77
USD
101846-096
Methyl-6-cyano-1H-indole-2-carboxylate
Methyl-6-cyano-1H-indole-2-carboxylate
Hazard: Irritant
Formula:
C₁₁H₈N₂O₂ MW: 200.1900 g/mol |
MDL Number:
MFCD07375393 CAS Number: 104291-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...