1-Isocyanato-4-(methylthio)benzene

Supplier: Matrix Scientific

038708-500MG
101846-044EA 144.9 USD
101846-044
1-Isocyanato-4-(methylthio)benzene
1-Isocyanato-4-(methylthio)benzene

Hazard: Irritant

Formula: C₈H₇NOS
MDL Number: MFCD00013878
CAS Number: 1632-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR