1-Fluoro-3-(isocyanatomethyl)benzene

Supplier: Matrix Scientific

Synonyms: 3-Fluorobenzyl isocyanate

038706-500MG
101846-042EA 217.35 USD
101846-042
1-Fluoro-3-(isocyanatomethyl)benzene
1-Fluoro-3-(isocyanatomethyl)benzene

Hazard: Irritant

Formula: C₈H₆FNO
MDL Number: MFCD02093692
CAS Number: 102422-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR