Ethyl 4-Fluorocinnamate

Supplier: Matrix Scientific
Warning

Synonyms: Ethyl (2E)-3-(4-fluorophenyl)acrylate4-Fluorocinnamic acid ethyl ester

038700-500MG
101846-030EA 87.4 USD
101846-030
Ethyl 4-Fluorocinnamate
Ethyl 4-Fluorocinnamate
Formula: C₁₁H₁₁FO₂
MW: 194.21 g/mol
MDL Number: MFCD02252625
CAS Number: 352-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR