Ethyl-4-(isocyanatomethyl)cyclohexanecarboxylate

Supplier: Matrix Scientific

038684-500MG
101846-002EA 217.35 USD
101846-002
Ethyl-4-(isocyanatomethyl)cyclohexanecarboxylate
Ethyl-4-(isocyanatomethyl)cyclohexanecarboxylate

Hazard: Irritant

Formula: C₁₁H₁₇NO₃
MDL Number: MFCD12027415
CAS Number: 945419-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR