1-(Isocyanatomethyl)-3-methoxybenzene

Supplier: Matrix Scientific

038683-500MG
101846-000EA 144.9 USD
101846-000
1-(Isocyanatomethyl)-3-methoxybenzene
1-(Isocyanatomethyl)-3-methoxybenzene

Hazard: Irritant

Formula: C₉H₉NO₂
MDL Number: MFCD03427184
CAS Number: 57198-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR