2-Isocyanato-1-methoxy-4-nitrobenzene

Supplier: Matrix Scientific

038670-500MG
101845-978EA 217.35 USD
101845-978
2-Isocyanato-1-methoxy-4-nitrobenzene
2-Isocyanato-1-methoxy-4-nitrobenzene

Hazard: Irritant

Formula: C₈H₆N₂O₄
MDL Number: MFCD00065712
CAS Number: 59741-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR