3-(4-Butoxyphenyl)-1H-pyrazole-4-carbaldehyde

Supplier: Matrix Scientific

038663-500MG
101845-962EA 217.35 USD
101845-962
3-(4-Butoxyphenyl)-1H-pyrazole-4-carbaldehyde
3-(4-Butoxyphenyl)-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₂
MDL Number: MFCD03834510
CAS Number: 696646-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR