3-(4-Isopropylphenyl)-1H-pyrazole-4-carbaldehyde

Supplier: Matrix Scientific

038662-500MG
101845-960EA 217.35 USD
101845-960
3-(4-Isopropylphenyl)-1H-pyrazole-4-carbaldehyde
3-(4-Isopropylphenyl)-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₄N₂O
MDL Number: MFCD03834509
CAS Number: 696646-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR